Would you like to log in again?
Accessories for TriggerGrip™ Dispenser, MicroCare
Supplier: MicroCare
MCC-BK3
76020-150CS 175.57 USD InStock
76020-150
Accessories for TriggerGrip™ Dispenser, MicroCare
Sprays
This device holds the both the aerosol can and the TriggerGrip™ dispenser neatly at the workbench; keeps the can from tipping over during use; ensures the aerosol can will empty completely.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty