Would you like to log in again?
CLEANER UNIV CONTACT PLASTIC-SAFE 10 OZ
Supplier: MicroCare
Universal Contact Cleaner 10oz^ Universal Contact Cleaner, Plastic-safe contact cleaner, Cleans mineral & silicone oils, dust, lint & grit, Dries in seconds with no residue, Cleans electronics & electrical equipment, Non-flammable, Compatible with Microcare TriggerGrip* cleaning system, Volume: 10oz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANER UNIV CONTACT PLASTIC-SAFE 10 OZ 76020-154 MCC-CCH10A Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty