Would you like to log in again?
CLEANER STENCIL/INK FLAMMABLE 5 GALLON
Supplier: MicroCare
Stencil cleaner 5 gallon^ Stencil cleaner, powerful fluid removes inks, UV curable inks, thick-film pastes, partially-cured epoxies. ScreenClean* reacts quickly with the contamination, breaking molecular bonds, speeding cleaning, Slow drying, Ozone-safe, Low GWP, Flammable, Volume: 5 gallon
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANER STENCIL/INK FLAMMABLE 5 GALLON 76020-156 MCC-CDIP Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty