Would you like to log in again?
WIPE PRESATURATED SCREENCLEAN 8IN X5IN
Supplier: MicroCare
ScreenClean* Presaturated Wipe 8x5in^ ScreenClean* Presaturated Wipe,Stencil, Squeegee and Misprint Cleaner, for removing inks, thick-film pastes and partially-cured epoxies, reacts quickly with the contamination, breaking molecular bonds, speeding cleaning and improving quality, Size: 8 in x 5 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WIPE PRESATURATED SCREENCLEAN 8IN X5IN 76020-159 MCC-CDIW Case of 1200 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty