Would you like to log in again?
REMOVER ALCOHOL-ENHNC FLUX PROCLEAN 12OZ
Supplier: MicroCare
ProClean* Alcohol-Enhanced Flux Remover 12oz^ ProClean* Alcohol-Enhanced Flux Remover, ESD-Safe, clear, liquid, Flash Point: 17 deg C, Flammability‎: Flammable, VOCs Content: 785g/L, Applications: Electronic Rework and Repair, For Use With: Triggergrip Cleaning System, Size: 12oz aerosol
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER ALCOHOL-ENHNC FLUX PROCLEAN 12OZ 76020-164 MCC-PRO Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty