Would you like to log in again?
REMOVER ALCOHOL-ENHNC FLUX PROCLEAN 1GAL
Supplier: MicroCare
ProClean* Alcohol-Enhanced Flux Remover 1gal^ ProClean* Alcohol-Enhanced Flux Remover, ESD-Safe, clear, liquid, Flash Point: 17 deg C, Flammability‎: Flammable, VOCs Content: 785g/L, Applications: Electronic Rework and Repair, For Use With: Triggergrip Cleaning System, Size: 1 gal mini pail
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER ALCOHOL-ENHNC FLUX PROCLEAN 1GAL 76020-166 MCC-PROG Case of 6 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty