Would you like to log in again?
CLEANER ALCOHOL FLUX PROCLEAN 5 GALLON
Supplier: MicroCare
ProClean* Cleaner 5 Gal^ ProClean* Cleaner for Rosin & No-Clean Fluxes & Pastes, Boiling Point: 77.6 deg C, Flash Point: 12 deg C, Odor: Alcohol, Low-cost cleaner for rosin, water based and no-clean fluxes, Use on solder pastes, fluxes and fresh chip bonder, Plastic-safe, Slow drying, Volume: 5 Gal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANER ALCOHOL FLUX PROCLEAN 5 GALLON 76020-170 MCC-PROP Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty