Would you like to log in again?
REMOVER HEAVY-DUTY FLUX SUPRCLEAN 1 GAL
Supplier: MicroCare
SuprClean* Heavy-Duty Rosin Flux Remover 1 gallon^ SuprClean* Heavy-Duty Rosin Flux Remover, Boiling Point: 41 deg C, Versatile both as degreaser & defluxer, Aggressive cleaning strength, Non-flammable, ESD-safe, Low Slight, Ether odor, Instant drying, No residues, no rinsing, Volume: 1gallon
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER HEAVY-DUTY FLUX SUPRCLEAN 1 GAL 76020-176 MCC-SPRG Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty