Would you like to log in again?
REMOVER UNIVERSAL RMA FLUX 10 OZ CAN
Supplier: MicroCare
Universal RMA Flux Remover 10 oz Can^ Universal RMA Flux Remover, Boiling Point: 19 C, Cleans rosin fuxes, Removes oil & grease, Fast drying, low odor, High purity, Non-flammable, Excellent silicone remover, Plastic safe, Compatible with Microcare TriggerGrip* cleaning system, Volume: 10 oz Can
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER UNIVERSAL RMA FLUX 10 OZ CAN 76020-184 MCC-UFR10A Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty