Would you like to log in again?
REMOVER VOC-FREE FLUX ULTRACLEAN 10OZ
Supplier: MicroCare
UltraClean* VOC-Free Flux Remover 10oz^ UltraClean* VOC-Free Flux Remover, Removes rosin-based fluxes, pastes, and silicone residues on circuit boards, Medium drying speed, Falmmable, Strongest non-chlorinated/halogen-free cleaner, TriggerGrip* cleaning system compatible, Plastic-safe, Volime: 10oz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER VOC-FREE FLUX ULTRACLEAN 10OZ 76020-188 MCC-VOC10A Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty