Would you like to log in again?
REMOVER NO CLEAN FLUX VERICLEAN (500 G)
Supplier: MicroCare
VeriClean* No Clean Flux Remover 1 Gallon^ VeriClean* No Clean Flux Remover, moderately aggressive circuit cleaning fluid, For light oils, grease and many adhesives, Dissolves silicone oils, coatings and adhesives quickly and safely, No residue / no rinsing, Plastic-safe, Volume: 1 Gallon
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER NO CLEAN FLUX VERICLEAN (500 G) 76020-190 MCC-DC1G Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty