Would you like to log in again?
REMOVER POLAR FLUX 10 OZ FOR LEAD-FREE
Supplier: MicroCare
Polar R RA RMA Flux Remover 10oz^ Polar R, RA, RMA Flux Remover, Dries residue free, Removes polar & ionic activators, Fast, extra-strength cleaning, minimizes white residuesHigh purity, Excellent silicone remover, Compatible with Microcare TriggerGrip* cleaning system, Volume: 10oz Can
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVER POLAR FLUX 10 OZ FOR LEAD-FREE 76020-232 MCC-PFR10A Case of 12 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty