Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proteinase K

Supplier: Zymo Research
D3001-2-5 D3001-2-20
76020-324EA 23.8 USD InStock
76020-324 76020-322
Proteinase K
Enzymes
Proteinase K is a stable serine protease with broad substrate specificity and will degrade many proteins in their native conformation, even in the presence of detergents (e.g., SDS).

  • Specifications: Lyophilized enzyme provided with Proteinase K Storage Buffer.
  • Enzyme Commission Number: (EC 3.4.21.64)
  • Source: Engyodontium album pH and Temperature Range: 4.0 to 12.0 (8.0 is optimum), 25 to 65°C.
  • Specific Activity: Over 30 units/mg protein
  • Unit Definition: One unit (U) of enzyme will hydrolyze urea-denatured hemoglobin to produce 1.0 μmole of tyrosine per minute at pH 7.5 at 37°C.

The enzyme is frequently used in molecular biology applications to digest unwanted proteins such as nucleases from DNA and/or RNA preparations from microorganisms, cells, and plants.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty