Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
DNA/RNA Shield™ Reagent, Zymo Research
Supplier: Zymo Research
Shield™
R1200-25 R1200-125 R1100-250 R1100-50
76020-418EA 433 USD InStock
76020-418 76020-440 76020-444 76020-420
DNA/RNA Shield™ Reagent, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.
  • Nucleic acid preservation (at ambient temperature; cold-free)
  • Pathogen inactivation (bacteria, fungus, parasites & viruses)
  • RNA Stability: Ambient temperature (4 to 25 °C) >1 month
  • DNA Stability: Ambient temperature (4 to 25 °C) >2 years
  • DNA and RNA: Frozen (<−20 °C): Indefinitely
DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialized equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty