Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water DNase-, RNase-free

Supplier: Zymo Research
Synonyms: Dihydrogen oxide

W1001-30 W1001-10 W1001-4 W1001-6 W1001-1
76020-504EA 17 USD InStock
76020-504 76020-502 76020-496 76020-500 76020-498
Water DNase-, RNase-free
Water


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty