Synonyms:
BCIG
, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
X1001-5
X1001-25
76020-508EA
16.4
USD
InStock
76020-508
76020-506
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 2% in DMF
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Sterile, ready to use X-Gal solution.
- Concentration: 2% w/v in DMF
- Storage: -20° C