Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ALPHA-FACTOR MATING PHEROMONE 240UL
Supplier: Zymo Research
Alpha-Factor Mating Pheromone 240ul^ Alpha-Factor Mating Pheromone, Aqueous solution, Concentration: 10mM in 0.1M sodium acetate pH5.2, Store at -20C, Purity: Minimum 98% (HPLC), Molecular Weight: 1684, Activity Test: Pass (G1 arrest testing), alter cell surface & nuclear determinants, Size: 240 ul
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ALPHA-FACTOR MATING PHEROMONE 240UL 76020-510 Y1001 Each (240µl) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty