Would you like to log in again?
RNA Clean & Concentrator™ Kits, Zymo Research
Supplier: Zymo Research
Clean & Concentrator™
R1080 R1016 R1019 R1017 R1018 R1015 R1013 R1014
76020-614EA 214.1 USD InStock
76020-614 76020-414 76020-612 76020-606 76020-604 76020-594 76020-608 76020-610
RNA Clean & Concentrator™ Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The RNA Clean & Concentrator™ (RCC™) kits facilitate the efficient removal of RNA polymerases, ligases, and RNA modifying enzymes.
  • Quick methods for cleaning and concentrating RNA
  • Zymo-Spin™ Column/Plate technology allows minimal elution volumes
  • Ideal for purification of RNA from aqueous phase following acid phenol extraction
The Zymo RNA Clean and Concentrator™ kits provide simple and reliable methods for the rapid preparation of high-quality RNA. The kit owes its simplicity to a unique single-buffer system and Zymo-Spin™ technology. Simply add the binding buffer to sample, adjust the conditions for binding by adding ethanol, then wash and elute the concentrated RNA. RNA ≥ 17 bases can be safely treated and recovered using these kits. The result is highly concentrated, purified RNA that is perfect for subsequent RNA-based methods, including RT-PCR, hybridization, and more.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty