Would you like to log in again?
AlphaTec® 68-4000 Chemical Barrier Coveralls, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaTec®
GR40-T-92-122-08 GR40-T-92-122-06 GR40-T-92-122-05 GR40-T-92-122-09 GR40-T-92-122-03 GR40-T-92-122-04 GR40-T-92-122-07 GR40-T-92-122-02
76045-302CS 598.89 USD InStock
76045-302 76020-690 76045-304 76020-694 76020-692 76020-698 76045-186 76020-696
AlphaTec® 68-4000 Chemical Barrier Coveralls, Ansell
Disposable Garments
Innovative multi-layer chemical barrier technology offering Type 3/4/5 protection.
  • Over 200 chemicals permeations tested, including chemical warfare agents
  • Innovative protective features include dual zip systems and double cuffs
  • Welded and taped seams, offering the highest barrier to liquids and particulates
  • Enhanced comfort; knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static
Engineered to provide an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
Certifications: AATCC76; ASTM F1671. CE Category III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5).
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty