Would you like to log in again?
ActivArmr® 97-125 Cut and Impact Protection Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
97125100
76020-744CS 3218.32 USD InStock
76020-744
ActivArmr® 97-125 Cut and Impact Protection Gloves, Ansell
Gloves
Certified impact protection for heavy-duty jobs with high cut risk.
  • Certified impact glove in accordance with EN 388:2016 impact requirements
  • Highly cut resistant, providing EN 388:2016 level C cut protection
  • Double-dip nitrile coating provides excellent grip and control when working with oils and other liquids
  • HPPE seamless knit glove keeps hands cool and comfortable
  • Double-dip nitrile coating defends against oil exposure and abrasion injuries
  • Designed for dynamic environments which have impact and cut hazards present
ActivArmr® 97-125 gloves have been designed for dynamic environments which have impact and cut hazards present. PVC bumpers and double-dip nitrile coating defends against oil exposure while providing ANSI Level 4 abrasion resistance and ANSI Level 3 Puncture resistance. The orange HIVIZ™ design increases awareness for user safety.
Certifications: Offers ANSI Level 4 Abrasion and ANSI 3 Puncture Protection.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty