Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BRDT Rabbit Polyclonal Antibody

Supplier: BioVision
AB286194-100UG
76020-776EA 896.34 USD InStock
76020-776
Anti-BRDT Rabbit Polyclonal Antibody
Antibodies
BrdT is similar to the RING3 protein family. It possesses 2 bromodomain motifs and a PEST sequence. The bromodomain is found in proteins that regulate transcription. BrdT drives a meiotic and post-meiotic gene expression program. It also controls the genome-wide post-meiotic genome reorganization that occurs after histone hyperacetylation in elongating spermatids. It may play a role in the transcriptional regulation of spermatogenesis. It also seems to have a structural ATP-independent role in the reorganization of acetylated chromatin.

Type: Primary
Antigen: BRDT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty