Would you like to log in again?
Red Blood Cell Lysis Buffer, 10X, BioVision Inc.
AB204733-100ML
76020-788EA 140.99 USD InStock
76020-788
Red Blood Cell Lysis Buffer, 10X, BioVision Inc.
Buffers Extraction Buffers
Buffer for efficient removal of RBCs from whole blood, tissues or tumor cells.
  • 10X solution provides 1L of RBC Lysis buffer in a convenient size.
  • Simple & convenient method for highly efficient removal of Red Blood cells from whole blood, tissues or tumor cells.
  • The reagent does not affect leukocytes, normal tissue, or tumor cells and does not interfere with down-stream functional assays.
Red Blood Cell Lysis buffer for quick removal of RBC from whole blood, tissues and tumor cells. 10X solution makes 1L of 1X final working buffer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty