Would you like to log in again?
Red Blood Cell Lysis Buffer, 1X, BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB286858-100ML
76020-790EA 81.77 USD InStock
76020-790
Red Blood Cell Lysis Buffer, 1X, BioVision
Buffers Extraction Buffers
Buffer for efficient removal of RBCs from whole blood, tissues or tumor cells.
  • Ready to use as supplied.
  • Simple & convenient method for highly efficient removal of Red Blood cells from whole blood, tissues or tumor cells.
  • The reagent does not affect leukocytes, normal tissue, or tumor cells and does not interfere with down-stream functional assays.
Red Blood Cell Lysis buffer for quick removal of RBC from whole blood, tissues and tumor cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty