Would you like to log in again?
Albumin (Albuminuria) Fluorometric Assay Kit , BioVision
Supplier: BioVision
AB241017-100TEST
76020-884EA 789.34 USD InStock
76020-884
Albumin (Albuminuria) Fluorometric Assay Kit , BioVision
Assays Protein Assays
Simple, Highly Sensitive Assay, HTS
  • Simple procedure, High-throughput suitable
  • Uses a specific probe (AB580) that specifically recognizes albumin The assay can detect as low as 20 mg/L of Albumin in samples.
Albuminuria is a pathological condition characterized by the presence of high concentrations of albumin in urine. Albumin concentration ranges between 30-300 mg/L in the patient’s urine sample, which is not detected by commercially available protein tests. Albuminuria has become a useful marker for the early detection of chronic kidney diseases in patients suffering from diabetes, hypertension, and cardiovascular diseases. BioVision’s Albumin (Albuminuria) Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects albumin concentrations in healthy and Albuminuric urine samples. This assay is based on the fluorometric detection of albumin using a probe (AB580) that specifically recognizes albumin (Ex/Em: 600/630 nm). AB580 does not cause fluorescence in presence of other proteins or metabolites such as IgG, urea, ascorbic acid and glucose. Our assay can detect albumin concentrations as low as 20 mg/L.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty