Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AMMC iodide ≥98% (by NMR)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: 3-[2-(N,N-diethyl-N-methylammonium)ethyl]-7-methoxy-4-methylcoumarin iodide

AB285378-25MG AB285378-5MG
76021-846EA 572.11 USD InStock
76021-846 76021-848
AMMC iodide ≥98% (by NMR)
AMMC iodide
AMMC iodide is a useful substrate for the human cytochrome P450 CYP2D6 isozyme. It is demethylated to the fluorescent product 3-[2-(N,N-diethyl-N-methyl ammonium)ethyl]-7-hydroxy-4-methylcoumarin (AMHC). After the AMMC is hydrolyzed, the pH is increased by addition of base for optimal fluorescence detection. AMMC has been used with human liver microsomes to determine CYP2D6 activity and inhibition.

Storage Temperature: -20°C


https://us-legacy.vwr.com/stibo/web/std.lang.all/23/44/23492344.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty