Would you like to log in again?
Cathepsin L Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB197012-100TEST
76022-134EA 1124.25 USD InStock
76022-134
Cathepsin L Inhibitor Screening Kit (Fluorometric), BioVision Inc.
Assays Enzyme Assays
Only kit on the market to screen Cathepsin L inhibitors
  • Convenient & Rapid
  • High-throughput adaptable assay to screen/study/characterize the potential inhibitors of Cathepsin L
  • Cathepsin L inhibitor, FF-FMK is included in the kit
Cathepsin L (CTSL, EC 3.4.22.15) is a lysosomal cysteine protease that is implicated in protein degradation, arthritis, apoptosis, and cancer. Biovision’s Cathepsin L Inhibitor Screening Kit utilizes the ability of active Cathepsin L to cleave the synthetic AFC-based peptide substrate to release AFC, which can be easily quantified using a fluorometer or fluorescence microplate reader. In the presence of a Cathepsin L inhibitor, the cleavage of this substrate is reduced/abolished resulting in decrease or total loss of the AFC fluorescence. This simple and high-throughput adaptable assay kit can be used to screen/study/characterize potential inhibitors of Cathepsin L.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty