Would you like to log in again?
Cell Surface Protein Isolation Kit, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB206998-10TEST
76022-148EA 877.51 USD InStock
76022-148
Cell Surface Protein Isolation Kit, BioVision
Assays Protein Assays
Simple and efficient method for isolation of cell surface proteins
  • Simple and efficient method for the isolation of cell surface proteins
  • Includes all required components for optimal labeling and isolation of the cell surface proteins
Cell surface proteins play a major role in signal transduction, cell adhesion, transport and serve as diagnostic and pharmacological targets. BioVision’s Cell Surface Protein Isolation Kit provides a simple and efficient method for the isolation of cell surface proteins. In this method, cells are first labeled with Sulfo-NHS-SS-Biotin, an amine-reactive, thiol-cleavable, biotinylation reagent. Cells are subsequently lysed and the labeled cell surface proteins are isolated using Streptavidin beads. The bound proteins are then released from beads by incubating with DTT solution. Biotinylation reagent is cell-membrane-impermeable with an extended spacer arm to reduce steric hindrances associated with streptavidin binding. This convenient kit provides all the required components for optimal labeling and isolation of the cell surface proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty