Would you like to log in again?
Thrombin Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB197007-100TEST
76022-158EA 1248.72 USD InStock
76022-158
Thrombin Inhibitor Screening Kit (Fluorometric), BioVision
Assays Protein Assays
Simple assay to screen thrombin inhibitors
  • Rapid & simple
  • Reliable test for screening potential inhibitors of thrombin
  • Includes thrombin Inhibitor [PPACK Dihydrochloride]
Thrombin enzyme (Factor IIa) is an important clotting factor that controls the transformation of soluble fibrinogen to insoluble active fibrin strands. Thrombin is a serine protease (EC 3.4.21.5) that catalyzes many coagulation-related reactions. Thrombin inhibitors are used as anticoagulants to prevent arterial and venous thrombosis. Some of these inhibitors are currently in clinical use while others are in clinical development. Biovision’s Thrombin Inhibitor Screening Kit utilizes the ability of Thrombin to cleave a synthetic AMC-based peptide substrate to release AMC, which can be detected by measuring its fluorescence at Ex/Em = 350/450 nm. In the presence of Thrombin specific inhibitors, the extent of cleavage reaction is reduced or completely abolished. The loss in the fluorescence intensity can be correlated to the amount of inhibitor present in the assay solution. The kit provides a simple and rapid method to screen potential inhibitors of Thrombin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty