Would you like to log in again?
CETP Activity Fluorometric Assay Kit II, BioVision
Supplier: BioVision
AB196995-100TEST
76022-208EA 1264.03 USD InStock
76022-208
CETP Activity Fluorometric Assay Kit II, BioVision
Assays Protein Assays
Convenient assay to measure CETP activity in animal serum, plasma or recombinant protein.
  • Simple, rapid & convenient assay to measure CETP activity in animal serum, plasma or recombinant protein.
  • Includes Inhibitor Control, Torcetrapib to validate the assay.
  • Comes with Positive Control (rabbit serum).
Cholesteryl ester transfer protein (CETP) is a plasma protein that transfers a cholesteryl ester from HDL to LDL or VLDL in exchange for a triglyceride. HDL plays an important role in lipid metabolism and cardiovascular health. HDL transports cholesterol to the liver for excretion or to steroidogenic tissues for steroid synthesis. HDL also plays an important role in the reverse cholesterol transport pathway, removing cholesterol from lipid-filled macrophages, protecting against atherosclerosis. Because of this function, CETP is viewed as a target to increase HDL, with CETP inhibition an active area of research and several CETP inhibitors at various stages of drug development. BioVision’s CETP Activity Fluorometric Assay Kit uses a self-quenched fluorescent neutral lipid that can be measured when transferred to an acceptor molecule. The fluorometric intensity is directly proportional to the amount of neutral lipid transfer. Rabbit serum is provided as a positive control and CETP inhibitor Torcetrapib is included for assay validation. This Assay Kit, in addition to measuring activity in serum, is also suitable for testing activity of recombinant protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty