Would you like to log in again?
VIAL W/LABEL/2 SCALE TEMP STRIP PP 90 ML
Supplier: CAPITOL VIAL INC.
Vial 90mL^ Vial, Twin Valve with Label and Dual Scale Temperature Strip, Material: Polypropylene, Cap type: Flip flop, Standard Twin Valve Seal, 48 mm diameter x 52 mm height, Capacity: 90mL / 3 ounce
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL W/LABEL/2 SCALE TEMP STRIP PP 90 ML 76022-516 10112TV Case of 400 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty