Would you like to log in again?
ZING Green Safety No Smoking Sign, Illinois
Supplier: ZING Green Safety Products
1848S 1850 1850S 2850S 1848D 2850 1850A 2850A
76023-032EA 19.21 USD InStock
76023-032 76023-022 76025-582 76023-030 76023-020 76025-584 76025-586 76023-028
ZING Green Safety No Smoking Sign, Illinois
Signs
The sign is available in 3 sizes, 7" x 5" (Decals and Labels only), 7" x 10", and 10" x 14", and four eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Self-Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Window Decals (face-adhesive signs) are designed to be placed on the inside of windows.
The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
Certifications: State No Smoking Laws and Regulations
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty