Would you like to log in again?
ZING Green Safety Eco Safety Projecting Sign, Spill Station
Supplier: ZING Green Safety Products
2605 2604
76024-120EA 42.3 USD InStock
76024-120 76024-118
ZING Green Safety Eco Safety Projecting Sign, Spill Station
Signs
Projecting sign is available as a V style or L style from recycled plastic.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Projecting V Sign clearly communicates critical safety information
Projecting L Sign clearly communicates critical safety information. The two-sided projecting design significantly increases visibility, and is ideal for designating critical workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 2.5"W x 7" projecting. Two mounting holes and adhesive tabs are included for easy installation.
The two-sided projecting design significantly increases visibility, and is ideal for designating important workplace locations. The sign is made in the USA from 70% post-consumer recycled plastic, and measures 7"H x 12"W x 5" projecting. Two mounting holes and adhesive tabs are included for easy installation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty