Would you like to log in again?
ZING Green Safety Eco Parking Sign Handicapped Parking Penalty Virginia
Supplier: ZING Green Safety Products
2705
76024-236EA 46.44 USD InStock
76024-236
ZING Green Safety Eco Parking Sign Handicapped Parking Penalty Virginia
Signs
Complies with state laws to designate access to handicap parking spaces.
  • 3M Engineer Grade Prismatic
  • Rated 7 years durability
  • High quality
ZING Eco Parking Sign "HANDICAPPED PARKING PENALTY" complies with Virginia state laws to designate access to handicap parking spaces. Sign is highly visible with 3M Engineer Grade Prismatic Reflective Sheeting and meets MUTCD Standards. Material is made from high quality 99% recycled .080 Aluminum (80% Post Consumer) that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Recovered materials also exceed EPA's recommended content levels for signage and are EPA CPG Listed. Recycled materials support sustainability initiatives and LEED Certified Building Projects. Sign measures 18"H x 12"W and includes pre-drilled holes for easy mounting. Made in the USA.
Certifications: State ADA Compliance, Meets MUTCD Standards
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty