Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZING Green Safety RecycLockout Lockout Tagout Station, 3 Padlock, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7113E 2726 7113
76026-242EA 158.96 USD InStock
76026-242 76024-246 76026-240
ZING Green Safety RecycLockout Lockout Tagout Station, 3 Padlock, ZING Enterprises
Security Locks
Available with Safety Locks or Aluminum Locks or Unstocked.

  • Can hold 6 padlocks
  • Can hold 4 lockout hasps
  • Can hold 3 safety tags with ties

ZING RecycLockout Lockout Tagout Station provides the flexibility you need to build a comprehensive Lockout Tagout program

Station boards have the flexibility to accommodate 1.5" or 3" shackle length padlocks and use unique hooks that allow two padlocks or hasps per hook. Padlocks can also be locked to the hooks for extra security. Lockout board is made from heavy-duty 99.9% recycled PETG that is break and impact resistant. Board and lockout devices are made from high content recycled materials that are UL Validated and support sustainability initiatives and LEED credits. Board measures 11"H x 11"W. Made in the USA Qty. 2 Padlocks, 1.5" shackle, keyed different with 1 key per padlock, color red

Qty. 1 Padlock, 3" shackle, keyed different with 1 key per padlock, color red

Qty. 1 Small Steel Hasp, 1" closed hole, accommodates up to 6 padlocks with shackles to 3/8" dia.

Qty. 3 Lockout Tags with legend DO NOT OPERATE and 50 lb. pull strength
Qty. 3 Cable Ties

Certifications: 29 CFR 1910.147

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty