Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZING Green Safety RecycLockout Lockout Tagout Station, 28 Padlock, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7116E 7116 2729
76026-254EA 454.31 USD InStock
76026-254 76024-252 76026-252
ZING Green Safety RecycLockout Lockout Tagout Station, 28 Padlock, ZING Enterprises
Security Locks
Available with Safety Locks or Aluminum Locks or Unstocked.

  • Capacity to hold 56 padlockS
  • 8 lockout hasps
  • 10 safety tags with ties

ZING Lockout/Tagout Station provides easy access to locks, hasps, tags and cable ties

Stations are designed to accommodate padlocks with a shackle length of 1.5” or 3", and the unique hooks can store two padlocks or hasps each. Padlocks can also be locked to the hooks for extra security. The station is made in the USA from recycled, impact-resistant plastic and is UL Validated for recycled content. The station measures 23"H x 23"W and includes 52 components. Station contents include:

Qty. 24 Padlocks, 1.5" shackle, keyed different with 1 key per padlock, color red

Qty. 4 Padlocks, 3" shackle, keyed different with 1 key per padlock, color red

Qty. 2 Large Steel Hasp, 1-1/2" closed hole, accommodates up to 6 padlocks with a shackle diameter up to 3/8"
Qty. 10 Lockout Tags with legend, “DO NOT OPERATE.” Tags have a 50 lb. pull strength.
Qty. 10 Cable Ties

Certifications: 29 CFR 1910.147

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty