Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZING Green Safety RecycLockout Lockout Tagout Kit, 32 Component, Deluxe Tool Box, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7129 2734
76024-262KT 390 USD InStock
76024-262 76026-270
ZING Green Safety RecycLockout Lockout Tagout Kit, 32 Component, Deluxe Tool Box, ZING Enterprises
Security Locks
ZING Lockout Tool Box provides a variety of lockout devices for on-the-job maintenance and repair operations.

  • Available with Safety Locks
  • Available with Aluminum Locks
  • High Quality

The tool box is made in the USA from durable, 100% recycled plastic, with open room to accommodate extra tools

The kit is UL Validated for recycled content, and supports sustainability initiatives and LEED credits. The tool box measures 13"W x 6.5"H x 5"D and includes 32 heavy-duty recycled lockout devices. Qty. 3 Breaker Lockouts, fits single, double and triple pole switches .3" - .6" high, .35" - .44" depth with or without a hole, accommodates 1 padlock with shackle diameter up to 9/32"

Qty. 1 Small Plug Lockout, universal fit closure accepting up to 1/2" dia. cord and 2"W x 3"L plugs, accommodates up to 3 padlocks with shackle diameter up to 9/16”

Qty. 1 Large Plug Lockout, universal fit closure accepting up to 1-1/4" dia. cord and 3"W x 5-1/2"L plugs, accommodates up to 3 padlocks with shackle diameter up to 9/16"

Qty. 3 Large Steel Hasps 1-1/2" closed hole, accommodates up to 6 padlocks with shackle diameter up to 3/8" dia.
Qty. 10 Lockout Tags with legend, “DO NOT OPERATE.” 50 lb. pull strength exceeds OSHA's requirement.
Qty. 10 Cable Ties

Certifications: 29 CFR 1910.147

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty