Would you like to log in again?
ZING Green Safety Eco Safety Sign, Exit, White on Red
Supplier: ZING Green Safety Products
2078A 2078 1078G 1078A 1078S 2078S 1078 2078G
76024-428EA 24.78 USD InStock
76024-428 76024-452 76024-430 76024-434 76024-456 76024-454 76024-432 76024-458
ZING Green Safety Eco Safety Sign, Exit, White on Red
Signs
The sign is available in two sizes, 7" x 10" and 10" x 14", and four eco-friendly materials.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Glow-in-the-Dark signs are manufactured from recycled plastic with a high-performance, safety grade photoluminescent coating. Signs will glow for a period of 10 hours on a full charge and require no electrical power source.
The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
Certifications: OSHA 1910.37,ASTM E2072 - 00
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty