Would you like to log in again?
ZING Green Safety Eco Safety Sign, DANGER Eye Protection
Supplier: ZING Green Safety Products
2097A 1097S 2097S 2097 1097 1097A
76024-470EA 29.62 USD InStock
76024-470 76022-716 76022-714 76023-514 76022-712 76023-512
ZING Green Safety Eco Safety Sign, DANGER Eye Protection
Signs
The sign is available in 2 sizes, 7" x 10", and 10" x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Certifications: OSHA 29 CFR 1910.145
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty