Would you like to log in again?
ZING Green Safety Eco Safety Sign, No Exit
Supplier: ZING Green Safety Products
2886A 1886S 1886A 1886G 2886 2886G 1886 2886S
76024-440EA 30.25 USD InStock
76024-440 76025-690 76024-444 76025-684 76024-442 76025-686 76025-688 76024-438
ZING Green Safety Eco Safety Sign, No Exit
Signs
Signs are available in 4 materials and 2 sizes; 7"Hx10"W and 10"Hx14"W.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
Glow-in-the-Dark signs are manufactured from recycled plastic with a high-performance, safety grade photoluminescent coating. Signs will glow for a period of 10 hours on a full charge and require no electrical power source.
The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
Certifications: NFPA 7.10.8.3.2
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty