Would you like to log in again?
ZING Green Safety RecycLockout Lockout Document Case, ZING Enterprises
Supplier: ZING Green Safety Products
6058
76025-966EA 172.8 USD InStock
76025-966
ZING Green Safety RecycLockout Lockout Document Case, ZING Enterprises
Security Locks
Stores important safety information and procedures to support compliance with lockout tagout standards.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Each case includes a built-in 3-ring holder that stores up to 300 sheets that can be conveniently carried to the job site or mounted to a wall. Case is highly visible with bright yellow/black colors and includes a lockable hasp on the door for extra security. Case is constructed from heavy gauge 75% recycled stainless steel that is UL Validated to support sustainability initiatives and LEED credits. Measures 12"H x 13"W x 3"D. Made in the USA.
Certifications: 29 CFR 1910.147
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty