Would you like to log in again?
ZING Green Safety RecycLock Safety Padlock, Keyed Different, 1-¹/₂" Shackle, 1-³/₄" Body, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7030 7044 7040 7038 7034 7032 7042 7036
76026-026EA 32.07 USD InStock
76026-026 76026-114 76026-110 76026-122 76026-034 76025-980 76026-030 76026-118
ZING Green Safety RecycLock Safety Padlock, Keyed Different, 1-¹/₂" Shackle, 1-³/₄" Body, ZING Enterprises
Security Locks
Available in red, yellow, blue, green, orange, purple, brown, or black.
  • Solid aluminum body
  • Steel shackle
  • Covered with plastic for corrosion resistance
ZING RecycLock Padlock is keyed different and includes one key that is unique and can only be used with that padlock. Padlock features a solid aluminum body with fully insulated protective covering for increased safety and corrosion resistance. Precision 6-pin cylinder has key-retaining and auto-locking functions. Lock is made from high quality 70% recycled materials that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Padlock contains no PVC and uses food industry approved lubricants. Lock body comes with trilingual label and measures 1-3/4" Length x 1-1/2" Wide. Steel shackle measures 1-1/2" Length x 1/4" dia.
Certifications: 29 CFR 1910.147
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty