Would you like to log in again?
Accredo Safety 1 Piece Steel Ball Valve Lockout, ZING Enterprises
Supplier: ZING Green Safety Products
7275 7274 7273
76026-090EA 36.37 USD InStock
76026-090 76026-094 76026-092
Accredo Safety 1 Piece Steel Ball Valve Lockout, ZING Enterprises
Security Locks
Convenient way to lockout ball valves.
  • High quality
  • Effective lockout device
  • Very durable
Small Steel Ball Valve Lockout locks most common ball valves in the OFF position
Large Steel Ball Valve Lockout locks most common ball valves in the OFF position. The maximum handle thickness is 25/64, and the maximum handle width is 1 31/32. The device features 15 holes for precise positioning of padlocks, with a maximum shackle diameter of 9/32.
Premier Small Ball Valve Lockout locks most common ball valves in the OFF position. The maximum handle thickness is ¾, and the maximum handle width is 1 ½. The device features 8 holes for precise positioning of padlocks, with a maximum shackle diameter of 3/8. The device is made from durable plastic (polyurethane).
The maximum handle thickness is 9/32, and the maximum handle width is 1 29/64. The device features 8 holes for precise positioning of padlocks, with a maximum shackle diameter of 9/32.
Certifications: OSHA 1910.147
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty