Would you like to log in again?
Accredo Safety Electrical Panel Lockout, ZING Enterprises
Supplier: ZING Green Safety Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7276 7279 7280 7255 7283 7281 7282 7278 7277
76026-348EA 25.99 USD InStock
76026-348 76026-106 76026-378 76026-104 76026-102 76026-100 76026-098 76026-096 76026-380
Accredo Safety Electrical Panel Lockout, ZING Enterprises
Security Locks
High quality lockout products.
  • High quality
  • Effective lockout device
  • Very durable
Electrical Panel Key Switch Lockout prevents keys from being inserted into switches while lockout is applied
The lockout accommodates 1 padlock with a maximum shackle diameter of ¼. The padlock is made from durable plastic and measures 3"Hx 3.5"Lx 3.5"W. Push Button/Switch Lockout provides a safe and effective method for locking out push buttons or switches on electrical panels. The device measures 2.5"H x 3"L x 3"W and accommodates 1 padlock, with a maximum shackle diameter of ¼. The device is made from durable, clear acrylic. The adhesive backing facilitates easy installation.
Push Button/Switch Lockout provides a safe and effective method for locking out push buttons or switches on electrical panels. The device measures 3"Hx3.5"Lx3.5"W and accommodates 1 padlock, with a maximum shackle diameter of ¼. The device is made from durable, clear acrylic. The adhesive backing facilitates easy installation.
Electrical Switch Lockout effectively locks out electrical switches. The device features durable, clear acrylic construction and measures 2.25"x2.25"x2.25"H. The device accommodates 2 padlocks with a maximum shackle diameter of ¼.
Electrical Switch Lockout, 45° angle cutout, effectively locks out electrical switches. The device features durable plastic construction and measures 3.1"x3.1"x2.25"H. The device accommodates 2 padlocks with a maximum shackle diameter of ¼. Adhesive strips facilitate easy installation.
Electrical Switch Lockout, 90° angle cutout, effectively locks out electrical switches. The device features durable plastic construction and measures 3.1"x3.1"x2.25"H. The device accommodates 2 padlocks with a maximum shackle diameter of ¼. Adhesive strips facilitate easy installation.
Extra Large Electrical Switch Lockout effectively locks out electrical switches. The device features durable plastic construction and measures 4.15"x4.15"x4.15"H. The device accommodates 2 padlocks with a maximum shackle diameter of ¼. Adhesive strips facilitate easy installation.
Round Electrical Switch Lockout effectively locks out electrical switches. The device measures 3"Hx3.5"Lx3.5"W and features durable, clear acrylic construction. The device is designed for use with switches to 2'' diameter.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty