Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZING Green Safety RecycLock Safety Padlock, Keyed Different, 3" Shackle, 1-³/₄" Body, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7048 7060 7054 7046 7056 7058 7050 7052
76026-138EA 36.2 USD InStock
76026-138 76026-126 76026-134 76026-146 76026-154 76026-130 76026-142 76026-150
ZING Green Safety RecycLock Safety Padlock, Keyed Different, 3" Shackle, 1-³/₄" Body, ZING Enterprises
Security Locks
Available in red, yellow, blue, green, orange, purple, brown, or black.

  • Solid aluminum body
  • Steel shackle
  • Covered with plastic for corrosion resistance

ZING RecycLock Padlock is keyed different and includes one key that is unique and can only be used with that padlock. Padlock features a solid aluminum body with fully insulated protective covering for increased safety and corrosion resistance. Precision 6-pin cylinder has key-retaining and auto-locking functions. Lock is made from high quality 70% recycled materials that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Padlock contains no PVC and uses food industry approved lubricants. Lock body comes with trilingual label and measures 1-3/4" Length x 1-1/2" Wide. Steel shackle measures 3" Length x 1/4" dia.

Certifications: 29 CFR 1910.147

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty