Would you like to log in again?
ZING Green Safety RecycLock Safety Padlock, Keyed Alike, 1-¹/₂" Shackle, 3" Long Body, ZING Enterprises
Supplier: ZING Green Safety Products
7083 7077 7085 7081 7079 7073 7071 7075
76026-168EA 35.56 USD InStock
76026-168 76026-176 76026-088 76026-188 76026-076 76026-172 76026-084 76026-080
ZING Green Safety RecycLock Safety Padlock, Keyed Alike, 1-¹/₂" Shackle, 3" Long Body, ZING Enterprises
Security Locks
Available in red, yellow, blue, green, orange, purple, brown, or black.
  • Solid aluminum body
  • Steel shackle
  • Covered with plastic for corrosion resistance
ZING RecycLock Padlock is keyed alike and includes one key that can be used with other padlocks within the same series. Padlock features a solid aluminum body with fully insulated protective covering for increased safety and corrosion resistance. Precision 6-pin cylinder has key-retaining and auto-locking functions. Lock is made from high quality 70% recycled materials that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Padlock contains no PVC and uses food industry approved lubricants. Lock body comes with trilingual label and measures 3" Length x 1-1/2" Wide. Steel shackle measures 1-1/2" Length x 1/4" dia.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty