Would you like to log in again?
ZING Green Safety RecycLockout Lockout Tagout Kit, 11 Component, General Application, ZING Enterprises
Supplier: ZING Green Safety Products
2731 7119
76026-260KT 207.5 USD InStock
76026-260 76024-256
ZING Green Safety RecycLockout Lockout Tagout Kit, 11 Component, General Application, ZING Enterprises
Security Locks
ZING Lockout Kit provides a variety of lockout devices for on-the-job maintenance and repair operations.
  • Available with Safety Locks
  • Available with Aluminum Locks
  • High Quality
The water-repellent and fire-retardant lockout pouch is made from heavy-duty, 100% recycled fabric with double-sewn seams and a PVC-free coating
The kit is UL Validated for recycled content and supports sustainability initiatives and LEED credits. The kit is made in the USA and measures 6"H x 7"W x 2"D. 11 Components are included. Qty. 2 Padlocks, 1.5" shackle, keyed different with 1 key per padlock, red
Qty. 1 Small Steel Hasp, 1" closed hole, accommodates up to 6 padlocks with a shackle diameter up to 3/8"
Qty. 1 Large Steel Hasp, 1-1/2" closed hole, accommodates up to 6 padlocks with a shackle diameter up to 3/8"
Qty. 3 Lockout Tags with legend, “DO NOT OPERATE.” 50 lb. pull strength exceeds OSHA’s requirements.
Qty. 3 Cable Ties
Certifications: 29 CFR 1910.147
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty