Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZING Green Safety RecycLockout Lockout Tagout Hasp, ZING Enterprises

Supplier: ZING Green Safety Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7106 7127 7102 7109 7108 7128 7107
76026-268EA 15.7 USD InStock
76026-268 76026-266 76026-234 76026-220 76026-232 76026-230 76026-228
ZING Green Safety RecycLockout Lockout Tagout Hasp, ZING Enterprises
Security Locks
All hasps are made from 100% recycled material.

  • UL Validated for high recycled content
  • High quality
  • Safety device

ZING RecycLockout Hasp/Tag Combo is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired

Steel hasps are offered with 1" or 1.5” closed hasp hole with pry resistant tabs for extra security. Hasp is high visible red and designed with 6 holes to accommodate padlocks with shackles up to 3/8" diameter. Lockout is made from heavy-duty 99.5% recycled steel that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measure 5"L x 1-1/2"W. Made in the USA.

Aluminum hasps are offered with 1" or 1.5” closed hasp hole that accommodates 6 padlocks with shackles up to 3/8" diameter. Hasp is high visible red and made from non-sparking 99.5% recycled aluminum that is UL Validated. Recycled materials support sustainability initiatives and LEED credits. Lockout measure 5"L x 1-1/2"W. Made in the USA.

Plastic hasps are offered with 1" or 1.5” closed hasp hole that accommodates 6 padlocks with shackles up to 3/8" diameter. Hasp is high visible red and made from non-conductive 50% recycled plastic that is UL Validated and supports sustainability initiatives and LEED credits. Lockout measure 5"L x 1-1/2"W.

Certifications: 29 CFR 1910.147

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty