Would you like to log in again?
ZING Green Safety RecycLockout Lockout Tagout Kit, 32 Component, Deluxe Tool Box, ZING Enterprises
Supplier: ZING Green Safety Products
7129 2734
76024-262KT 390 USD InStock
76024-262 76026-270
ZING Green Safety RecycLockout Lockout Tagout Kit, 32 Component, Deluxe Tool Box, ZING Enterprises
Security Locks
ZING Lockout Tool Box provides a variety of lockout devices for on-the-job maintenance and repair operations.
  • Available with Safety Locks
  • Available with Aluminum Locks
  • High Quality
The tool box is made in the USA from durable, 100% recycled plastic, with open room to accommodate extra tools
The kit is UL Validated for recycled content, and supports sustainability initiatives and LEED credits. The tool box measures 13"W x 6.5"H x 5"D and includes 32 heavy-duty recycled lockout devices. Qty. 3 Breaker Lockouts, fits single, double and triple pole switches .3" - .6" high, .35" - .44" depth with or without a hole, accommodates 1 padlock with shackle diameter up to 9/32"
Qty. 1 Small Plug Lockout, universal fit closure accepting up to 1/2" dia. cord and 2"W x 3"L plugs, accommodates up to 3 padlocks with shackle diameter up to 9/16”
Qty. 1 Large Plug Lockout, universal fit closure accepting up to 1-1/4" dia. cord and 3"W x 5-1/2"L plugs, accommodates up to 3 padlocks with shackle diameter up to 9/16"
Qty. 3 Large Steel Hasps 1-1/2" closed hole, accommodates up to 6 padlocks with shackle diameter up to 3/8" dia.
Qty. 10 Lockout Tags with legend, “DO NOT OPERATE.” 50 lb. pull strength exceeds OSHA's requirement.
Qty. 10 Cable Ties
Certifications: 29 CFR 1910.147
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty