Would you like to log in again?
ZING Green Safety Safety Padlock, Keyed Different, 3" Shackle, 1-⁹/₁₆" Body, ZING Enterprises
Supplier: ZING Green Safety Products
7204 7201 7200 7203 7202
76026-288EA 47.47 USD InStock
76026-288 76026-286 76026-284 76026-282 76026-290
ZING Green Safety Safety Padlock, Keyed Different, 3" Shackle, 1-⁹/₁₆" Body, ZING Enterprises
Security Locks
Available in red, yellow, blue, green, or black.
  • Solid aluminum body
  • Steel shackle
  • Covered with plastic for corrosion resistance
ZING Aluminum Padlocks are designed to support OSHA Lockout requirements and prevent energization of equipment that could harm employees during servicing. Padlock body is made of lightweight premium aluminum that is non-sparking and non-magnetic and shackle is made from hardened alloy steel with protective plating for extreme corrosion resistance. Padlock uses high content recycled material that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others. Padlocks are keyed different and include two keys per lock. Padlocks feature precision 6-pin cylinder for maximum key difference. Lock body measures 1-9/16” H x 1-1/2”W x 3/4”D, Shackle length is 3".
Certifications: 29 CFR 1910.147
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty