Would you like to log in again?
ZING Green Safety Eco Beard/Hair Net Dispenser, Clear with Hinged Lid
Supplier: ZING Green Safety Products
7306
76026-314EA 165.85 USD InStock
76026-314
ZING Green Safety Eco Beard/Hair Net Dispenser, Clear with Hinged Lid
Signs
High quality dispenser.
  • Made from 100% recycled PETG
  • Superior durability
  • Crack resistance over other materials like acrylic
ZING Beard/Hair Net Dispenser conveniently stores beard or hair nets for quick and easy access. Beard/Hair Net Dispenser is made from heavy-duty 100% recycled PETG providing superior durability and crack resistance over other materials like acrylic. Dispenser is ideal for high use applications and can be mounted on a wall or placed on a counter-top. Durable PETG hinges support a lid that keeps contents clean and dust free. Recycled materials support sustainability initiatives and LEED Credits. Dispenser uses clear plastic and includes pre-drilled holes for easy mounting. Measures 9"H x 7.5"W x 8"D. Beard/Hair Nets Not Included. Made in the USA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty